missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 2% (w/v) with Mixed Indicator, Ricca Chemical

For Analysis of Ammonia and Nitrogen (Kjeldahl)

$126.78 - $397.00

Chemical Identifiers

Molecular Formula BH3O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
106416
View Documents Promotion Details
Ricca Chemical Company
106416
500 mL Natural Poly Bottle
Each for $126.78
Only null left
 
1064-32
View Documents Promotion Details
Ricca Chemical Company
106432
1 L Natural Poly Bottle
Each for $211.56
Only null left
 
1064-1
View Documents Promotion Details
Ricca Chemical Company
10641
4 L Natural Poly Bottle
Each for $397.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Mixture
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid

Specifications

0.0016,7.0E-4,95.0
Blue
Liquid
BH3O3
Natural Poly Bottle
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
Laboratory
Boric Acid
1.58,7.0E-4,0.0016
64-17-5,7220-79-3,10043-35-3,7732-18-5,845-10-3
500 mL
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
OB(O)O
Mixture
CHEBI:33118
2% (w/v)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'