missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 2% (w/v) with Mixed Indicator, Ricca Chemical

Catalog No. 106432
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
106432 1 L Natural Poly Bottle
106416 500 mL Natural Poly Bottle
10641 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 106432 Supplier Ricca Chemical Company Supplier No. 106432

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Analysis of Ammonia and Nitrogen (Kjeldahl)

Chemical Identifiers

Molecular Formula BH3O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Min % 0.0016, 7.0E-4, 95.0
Color Blue
CAS Max % 1.58, 7.0E-4, 0.0016
CAS 64-17-5, 7220-79-3, 10043-35-3, 7732-18-5, 845-10-3
Physical Form Liquid
Quantity 1 L
Packaging Natural Poly Bottle
Grade Laboratory
Concentration or Composition (by Analyte or Components) 2% (w/v)
Chemical Name or Material Boric Acid