missing translation for 'onlineSavingsMsg'
Learn More

Borax, For Silica, 1% (w/v), LabChem™

Catalog No. lc117101
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC117101 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC117101 Supplier LabChem Supplier No. LC117101
Only null left

Chemical Identifiers

CAS 1303-96-4 , 7732-18-5
Synonym Sodium Borate; Sodium Tetraborate

Specifications

Color Colorless
CAS Max % 99, 1
Linear Formula Na2B4O7·10H2O
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 381.37
Percent Purity 1% (w/v)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Boron
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Borax, For Silica
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature