missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane - Morpholine Complex 95.0+%, TCI America™
$91.33 - $247.92
Chemical Identifiers
| CAS | 4856-95-5 |
|---|---|
| Molecular Formula | C4H12BNO |
| Molecular Weight (g/mol) | 100.96 |
| MDL Number | MFCD00066736 |
| InChI Key | IFVAHVOGOBRFSP-UHFFFAOYSA-N |
| Synonym | morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine |
| PubChem CID | 6327109 |
| IUPAC Name | morpholine borane |
| SMILES | B.C1COCCN1 |
Chemical Identifiers
| 4856-95-5 | |
| 100.96 | |
| IFVAHVOGOBRFSP-UHFFFAOYSA-N | |
| 6327109 | |
| B.C1COCCN1 |
| C4H12BNO | |
| MFCD00066736 | |
| morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine | |
| morpholine borane |
Specifications
| 4856-95-5 | |
| White-Yellow | |
| MFCD00066736 | |
| 2921 | |
| IFVAHVOGOBRFSP-UHFFFAOYSA-N | |
| morpholine borane | |
| 6327109 | |
| ≥95.0% (T) | |
| Borane - Morpholine Complex |
| 97°C | |
| C4H12BNO | |
| 5 g | |
| morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine | |
| B.C1COCCN1 | |
| 100.96 | |
| 100.96 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : QD8530000
TSCA : Yes