missing translation for 'onlineSavingsMsg'
Learn More

Borane - Morpholine Complex 95.0+%, TCI America™

Supplier:  TCI America M089825G

 View more versions of this product

Catalog No. M089825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Borane - Morpholine Complex
97°C
C4H12BNO
25 g
morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine
B.C1COCCN1
100.96
100.96
Crystalline Powder
4856-95-5
White-Yellow
MFCD00066736
2921
IFVAHVOGOBRFSP-UHFFFAOYSA-N
morpholine borane
6327109
≥95.0% (T)

Chemical Identifiers

4856-95-5
100.96
IFVAHVOGOBRFSP-UHFFFAOYSA-N
6327109
B.C1COCCN1
C4H12BNO
MFCD00066736
morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine
morpholine borane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QD8530000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions