accessibility menu, dialog, popup

UserName

ONHBH3

CAS RN 4856-95-5

IUPAC Name: morpholine borane
Synonyms: morpholine borane
Molecular Weight (g/mol): 100.96
Molecular Formula: C4H12BNO
InChi Key: IFVAHVOGOBRFSP-UHFFFAOYSA-N
SMILES: B.C1COCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results