missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bond-Breaker™ TCEP Solution, Neutral pH

Catalog No. PI77720
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77720 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI77720 Supplier Thermo Scientific™ Supplier No. 77720
  • ONLINE EXCLUSIVE PRICE
    $502.35
    $459.44 / Each

    Save $42.91

    9.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficiently reduce protein disulfide bonds with this stable, odorless, 0.5M solution of TCEP reducing agent that is ideal for making reducing SDS-PAGE sample buffers.

Thermo Scientific™ Bond-Breaker TCEP Solution, Neutral pH is a stable, 0.5M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulfide bonds.

Highlights:

  • Ready-to-use, odorless, stable and neutral 0.5M TCEP solution
  • Eliminates TCEP·HCl stock solution preparation and neutralization prior to SDS-PAGE gel loading
  • Convenient, time-saving format for TCEP reductions
  • Neutral pH minimizes possibility of amide bond cleavage during reduction
  • Room temperature-stable, saves valuable refrigerator space
  • Contributes to more pleasant, safer laboratory environment

Specifications

Content And Storage Store at room temperature.
Product Line Bond-Breaker
Form Liquid
Quantity 5 mL
Product Type Disulfide Reducing Reagent
Can other reducing agents other than DTT or BME be used to reduce proteins prior to electrophoresis? For example, what about TCEP (Tris Carboxy Ethyl Phosphene)?

TCEP, Tris Carboxy Ethyl Phosphene is an alternative sulfhydryl reducing agent for protein samples. It is an extremely potent and effective reducing agent for particularly ‘difficult' proteins. It is compatible with the Tris-Glycine gels and NuPAGE gels. It should be added to the sample buffer for these systems. 20 mM final (maximum) concentration is sufficient for samples. You may add alkylating agents, e.g. Iodine (50 mM Iodoacetic acid), to prevent re-forming of S-S bonds but it is not necessary. Do not heat because this will hydrolyze much of your sample. Instead let the sample sit for several minutes at RT and then load.

For Research Use Only. Not for use in diagnostic procedures.