missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Bis-Tris Plus Resupply Pack, 4 to 12%, WedgeWell™ format

Catalog No. NW0412CRS10
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
NW0412CRS10 15-well
NW0412ARS10 10-well
NW0412BRS10 12-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NW0412CRS10 Supplier Invitrogen™ Supplier No. NW0412CRS10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Bolt Bis-Tris Plus Resupply Pack optimizes your protein separation workflow. This pack includes precast gels, running buffer, and protein ladders, enough for 100 gel runs.

The Bolt Bis-Tris Plus Resupply Pack optimizes your protein separation workflow. This pack includes precast gels, running buffer, and protein ladders, enough for 100 gel runs. Bolt Bis-Tris Plus gels offer the fastest Bis-Tris gel run time, superior broad-range separation, and maintain protein integrity due to their neutral pH environment. Designed for optimal separation of diverse protein sizes under denaturing conditions, they are ideal for western blot transfer and analysis. With a long 16 month shelf life and the innovative WedgeWell format for easier sample loading, this pack is a reliable and convenient solution for your laboratory needs.

The Bolt Bis-Tris Plus Resupply Pack contains:

  • Bolt Bis-Tris Plus gels, WedgeWell format (10 boxes, 100 gels)
  • Bolt MES Running Buffer for 100 gel runs, 20X (4 bottles, Cat. No. B0002)
  • PageRuler Plus Pre-stained Protein Ladder for 100 gel runs, 10 to 250 kDa (Cat. No. 26619)

About Bolt Bis-Tris Plus Gels

Bolt Bis-Tris Plus gels are high-performance precast polyacrylamide gels designed to provide optimal broad range separation of proteins under denaturing conditions. These gels operate at a neutral pH, minimizing protein modifications and ensuring protein integrity, making them ideal for western blot transfer and analysis. The unique WedgeWell design allows for up to 2X more sample volume loading than conventional precast gels, facilitating detection of proteins in extremely diluted samples or visualization of low-abundance proteins. Bolt Bis-Tris Plus Gels, combined with Bolt LDS Sample Buffer and the Invitrogen NuPAGE Antioxidant running buffer additive, deliver sharp, clear protein bands, preserving protein integrity throughout electrophoresis. Stable at room temperature, these gels are a practical and efficient choice for your lab.

Separation

For running Invitrogen protein gels, you have the flexibility to use either the Mini Gel Tank (A25977) or the XCell SureLock Mini-Cell (EI0001). Both systems are designed to provide optimal performance. The Mini Gel Tank's unique side-by-side design offers efficient and convenient electrophoresis for both precast and SureCast handcast mini gels. Alternatively, the XCell SureLock Mini-Cell allows quick, easy, and leak-free running of mini gels, without the need for any clamps. This compatibility gives you the freedom to choose the best tool for your specific lab and experiment requirements. Additionally, the Mini Gel Tank is conveniently available through Invitrogen Precast Gel Welcome Packs along with both sample buffer and reducing agent, providing an all-in-one solution for your electrophoresis needs.

Transfer

For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Specifications

Content And Storage • Bolt Bis-Tris Plus gels, 10 x 10 gels
• Bolt MES Running Buffer (20X), 4 bottles
• PageRuler Plus Pre-stained Protein Ladder (10 to 250 kDa)
Gel Percentage 4 to 12%
Gel Size Mini
Gel Type Bis-Tris Plus
Separation of Protein
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 15-well
Quantity 1 Kit
For Use With (Equipment) Mini Gel Tank

For Research Use Only. Not for use in diagnostic procedures.