missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format

Catalog No. NW00120BOX
Encompass
Change view
Click to view available options
Wells:
10-well
12-well
15-well
17-well
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
NW00120BOX 10-well
NW00122BOX 12-well
NW00125BOX 15-well
NW00127BOX 17-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NW00120BOX Supplier Invitrogen™ Supplier No. NW00120BOX

Requires

Mini Gel Tank (Mfr. No. A25977)

Invitrogen Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of a wide range of protein sizes under denaturing conditions. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial.

Invitrogen Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of a wide range of protein sizes under denaturing conditions. Similar to NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications with increased loading capacity. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial.

Use Bolt Bis-Tris Plus gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four gel types and multiple well formats. Each box contains 10 gels. A Mini Gel Tank (A25977) is required for use of all Bolt gels.

Features include:
Optimized run conditions—separate your proteins using constant voltage in approximately 20 minutes
Long shelf life—store gels for at least 16 months at 4–25°C
Better protein integrity—neutral pH sample preparation process preserves your proteins
Wedge-shaped wells—for easier sample loading and up to two times higher sample volume

Choose the right Bolt Bis-Tris Plus gels for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES SDS (B000202) or Bolt MOPS SDS (B000102) running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.

Convenient to use
Bolt Bis-Tris Plus gels can be run using constant voltage in approximately 20 minutes with the Bolt MES running buffer. Bolt Bis-Tris Plus gels offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than Tris-glycine–based gels. In addition, Bolt Bis-Tris Plus gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier. And Bolt Bis-Tris Plus gels have a shelf life of at least 16 months at 4°C to 25°C.

For transferring your proteins to a membrane, we recommend using for Bolt Transfer buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001) .

Specifications

Gel Percentage 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris Plus
Recommended Applications Denaturing
Sample Loading Volume up to 60 μL
Separation Range 3.5 to 80 kDa
Separation Type Denaturing
Wells 10-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Room Temperature
Storage Requirements Store at 4–25°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line Bolt, WedgeWell
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less
Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

What are the storage conditions for Bolt gels?

Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

How much methanol is required per transfer when using the Mini Blot Module and Bolt Bis-Tris Plus gels?

We recommend using 1x Bolt Transfer buffer with 10% methanol.

How many sponge pads do you recommend using for transfer of Bolt Bis-Tris Plus gels using the Mini Blot Module?

We recommend using 2 sponge pads to start with. If the sponge pads become thinner with extended use, then an additional sponge pad can be used.

Is there a wet transfer blot module available for the Mini Gel Tank? Are Bolt Bis-Tris Plus gels compatible with other transfer methods?

Yes, we offer the Mini Blot Module (Cat. No. B1000), designed to be used with the Mini Gel Tank. This blot module will also work with the Bolt Mini Gel Tank (discontinued as of December 31, 2014). Please note that the Bolt Mini Blot Module (discontinued as of December 31, 2014) is also compatible with both the Bolt Mini Gel Tank and the Mini Gel Tank.

Bolt Bis-Tris Plus gels can also be transferred using the XCell SureLock Mini Cell, iBlot Dry transfer system, or using the Invitrogen Semi-Dry Blotter.

My gel run is faster than normal with poor resolution. What could be causing this problem?

Here are possible causes and solutions:

- Buffers are too concentrated or incorrect. Check buffer recipe; dilute or re-make if necessary.
- Voltage, current or wattage is set at a higher limit. Decrease power conditions to recommended running conditions.

My gel will not run. Can you please help me troubleshoot?

It is important to determine whether the problem is with the power supply, the apparatus or the gel. Often, it helps to switch out the power supply or the lid to see if there is a faulty contact. Also, check to see whether the tape from the bottom of the gel cassette has been removed and whether the buffer core is damaged. Additionally, make sure there is sufficient buffer in the electrophoresis tank to cover the wells of the gel.

Which of your protein gels can I run using the Mini Gel Tank?

Our New Bolt Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Invitrogen Mini gels and NuPAGE Mini gels can be run using the Mini Gel Tank. Please note that our original Bolt Bis-Tris Plus Mini gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) can only be run in the Bolt Mini Gel Tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).

Can I run the Bolt Bis-Tris Plus gels in the XCell SureLock Mini-Cell?

Our Original Bolt Bis-Tris Plus gels (Cat. No. BGxxxxxBOX), discontinued as of December 31, 2014) do not fit in the XCell SureLock Mini-Cell. They need to be run in the Bolt Mini Gel Tank (discontinued as of December 31, 2014). Our New Bolt Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX) can be run using the XCell SureLock Mini-Cell (or Mini Gel Tank, Cat. No. A25977).

Which of your protein gels can I run using the XCell SureLock Mini-Cell?

Our New Bolt Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Invitrogen Mini gels and NuPAGE Mini gels can be run using the XCell SureLock Mini-Cell.

Which gel chamber should I use to run your Invitrogen Mini and Midi protein gels?

Our Original Bolt Bis-Tris Plus Mini gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) can only be run in the Bolt Mini Gel Tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).

Our New Bolt Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Invitrogen Mini gels and NuPAGE Mini gels can be run using the Mini Gel Tank, or XCell SureLock Mini-Cell. To run these gels using the Bolt Mini Gel Tank (discontinued as of December 31, 2014), upgrading of the tank is necessary by replacing the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732, Cassette Clamp Cam Handle Set). Instructions for replacement of the cam handles can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gel-electrophoresis-chamber-systems/mini-gel-tank/resources-upgrading-bolt-mini-gel-tank.html).

Our Midi gels can be run using the XCell4 SureLock Midi-Cell.

After western detection, my membrane has a lot of spots. What could have gone wrong?

Here are possible causes and solutions:

- Membrane blotting pads are dirty or contaminated. Soak pads with detergent and rinse thoroughly with purified water before use. Replace pads when they become worn or discolored.
- Blocking was uneven. The incubation dish must be sufficiently big to allow thorough coverage of membrane. Shake or agitate during each step.

I am getting a lot of non-specific binding after western detection. Can you offer some tips?

Here are possible causes and solutions:

- Membrane contaminated by fingerprints or keratin proteins: Wear clean gloves at all times and use forceps when handling membranes. Always handle membranes around the edges.
- Concentrated secondary antibody used: Make sure the secondary antibody is diluted as recommended. If the background remains high, but with strong band intensity, decrease the concentration of the secondary antibody.
- Concentrated Primary antibody used: Decrease the concentration of the primary antibody.
- Affinity of the primary antibody for the protein standards: Check with the protein standard manufacturer for homologies with primary antibody.
- Insufficient removal of SDS or weakly bound proteins from membrane after blotting: Follow instructions for membrane preparation before immunodetection.
- Short blocking time or long washing time: Make sure that each step is performed for the specified amount of time.

I am getting very high background after western detection. Can you please offer some tips?

Here are possible causes and solutions:

- Insufficient blocking or non-specific binding: We suggest trying our WesternBreeze Blocker/Diluent (Cat. No. WB7050).
- Membrane is contaminated: Use only clean, new membranes. Wear clean gloves at all times and use forceps when handling membranes.
- Higher intrinsic background with PVDF membranes: Switch to nitrocellulose membranes.
- Nitrocellulose membrane not completely wetted: Follow instructions for pre-wetting the membrane.
- Blot is overdeveloped: Follow recommended developing time and remove blot from substrate when signal - to -noise ratio is acceptable.
- Insufficient washing ; Follow recommended number of washes. In some cases, it may be necessary to increase the number or duration of washes.
- Concentrated secondary antibody used: Determine optimal antibody concentration by performing a dot blot and dilute antibody as necessary.
- Concentrated primary antibody used: Determine optimal antibody concentration by performing a dot blot and dilute antibody as necessary.

I am unable to visualize my protein bands after western detection. What is the problem?

Here are possible causes and solutions:

- The primary antibody and secondary antibody are not compatible: Use a secondary antibody that was raised against the species in which the primary antibody was raised.
- The primary antibody is too dilute: 1) Use a more concentrated antibody solution. 2) Incubate longer (e.g., overnight) at 4 degrees C. 3) Use fresh antibody and keep in mind that each time an antibody solution is used, its effective antibody concentration decreases.
- Something in your blocking buffer interferes with binding of the primary and/or secondary antibody: Try an alternate blocking buffer ± a mild surfactant like Tween-20 (0.01-0.05% v/v). There are many blocking buffer recipes available, based on non-fat dry milk, BSA, normal serum, gelatin and mixtures of these and other materials. Note that BSA (1-5%) is considered the best blocker for nitrocellulose membranes. It is easy to check the efficacy of different blocking buffers by performing dot-blots.
- The primary antibody does not recognize the protein in the species being tested: 1) Evaluate primary antibodies by dot-blotting first to how well they react with your protein. 2) Check the immunogen sequence, if provided, and determine if it is found in your protein. 3) If no immunogen sequence is available, perform a PubMed/BLAST alignment to assess the degree of homology between your target protein and the protein against which the antibody was generated. Note that many antibodies against human proteins will also recognize the non-human primate version because there is usually a high degree of amino acid identity. In contrast, many antibodies against human proteins will not recognize the corresponding proteins from rodents (and vice versa). Remember that significant homology between sequences does not guarantee that the antibody will recognize your protein. 4) Always run the recommended positive control, if available.
- Insufficient protein is bound to the membrane or the protein of interest is not abundant enough in the sample: 1) Load at least 20-30 ?g protein per lane on your gels (as a starting point), since proteins representing less than ~0.2% of the total protein are difficult to detect on western blots. 2) Use an enrichment step to increase the concentration of the target protein. For example, prepare two nuclear lysates prior to blotting nuclear proteins or perform an immunoprecipitation (IP) prior to SDS-PAGE. 3) Reduce the volume of cell extraction buffer used to lyse your cells or tissue. 4) Be sure to use freshly prepared protease inhibitors and phosphatase inhibitors, if needed, in your protein extraction buffer. 5) Run the recommended positive control, if available.
- Poor or no transfer of the proteins to the membrane 1) Check the protein transfer efficiency with a reversible protein stain like Invitrogen Reversible Membrane Protein Stain, ponceau S, amido black or use pre-stained molecular weight standards. 2) Verify that the transfer was performed with the correct electrical polarity. 3) Remember that proteins with basic pI values (e.g., histones) and high MW may not transfer well. 4) Remember that if your target protein has a low MW (≤10 kDa), it may transfer more quickly than expected. 5) If you are using PVDF membranes, make sure to pre-soak the membrane in methanol first before soaking it in transfer buffer. Note that methanol in transfer buffer increases protein binding to nitrocellulose, but omitting methanol can increase transfer efficiency of high MW proteins. 6) Low MW proteins may pass through the 0.45 µm pores in nitrocellulose membranes, so switch to NC with 0.2 or 0.1 µm pores instead.
- Excessive washing or blocking of the membrane:- 1) Avoid over-washing the membrane. Extra washing will not allow you to visualize your protein of interest if there are other problems with your blot. 2) Avoid over-blocking by using high concentrations of the blocking buffer components or long incubation times. Too much blocking can prevent your antibodies from binding to your protein. Gelatin, in particular, can mask proteins on the blot, so avoid it, if possible. Milk can also mask proteins, so instead of using 5% milk in your blocking buffer, try using it at 0.5% instead, or remove it altogether. 3) Switch to a different blocking reagent and/or block the blot for less time.
- Using the same solution of diluted primary antibody repeatedly: Use freshly-diluted antibody for each western blot because the effective concentration of a diluted antibody decreases each time it is re-used. Also, remember that dilute solutions of antibodies are less stable and may lose their activity rapidly.
- The enzyme conjugated to your secondary antibody is not working: 1) Make a fresh dilution of your secondary antibody conjugate each time you need it. Enzymes (and antibodies) may lose activity quickly in dilute solutions. 2) Omit sodium azide in buffers if you are using HRP-conjugated antibodies. 3) Avoid high heme concentrations (from blood contamination), which can interfere with HRP-based detection. 4) Avoid using phosphate in buffers with alkaline phosphatase-antibody conjugates because phosphate inhibits enzyme activity.
- Your colorimetric or other detection reagent is old and inactive: 1) Use fresh enzyme substrate for each experiment. 2) Don't use ready-to-use substrate reagents if they have changed color on their own or if they have passed their expiration date. 3) Do not dilute substrate solutions unless instructed to do so in the product manual.

I ran my protein sample on one of your gels and the bands look non-distinct and smeary after western detection. What should I do?

Here are some suggestions:

- Make sure that the correct amount of protein is loaded per lane; loading too much protein can cause smearing.
- Bands will not be as well resolved in low percentage gels; try using a higher percentage gel.
- This may be due to the antibody being too concentrated. We recommend following the manufacturer's recommended dilution or determining the optimal antibody concentration

I ran my protein samples on one of your protein gels and saw V-shaped protein bands. Can you please help me troubleshoot?

V-shaped protein bands are caused by the presence of DNA in the sample. The artifact might be eliminated or minimized by shearing the DNA with additional sonication after the SDS-solubilization step. Alternatively, the DNA can be removed from the sample using an ultra-centrifuge.

I am getting wavy dye fronts when using Bolt Bis-Tris Plus gels with the Bolt Mini Gel Tank. Why is this?

Here are some causes and solutions for wavy dye fronts:

1) Difference in buffer level between the inner and outer buffer chambers: Both buffer chambers must be filled up to the electrode with wells completely covered. This will not only prevent leaks from the inside to the outside but will also act a heat sink and prevent wavy dye fronts.
2) Using running buffer that was diluted more than 1X: We recommend using 1X running buffer.
3) Using old running buffer: Make sure that the running buffer is fresh and don't reuse the running buffer.

My Bolt Bis-Tris Plus gel is running backwards but if I reverse the leads on the Bolt Mini Gel Tank, it runs correctly. Can you please help me troubleshoot?

It is likely that the Bolt gel cassette was inserted backwards into the unit (large plate facing the front and the wells facing the back) even though this is pretty difficult to do. When the gel is inserted backwards, the current flows from the bottom of the gel to the top, resulting in the samples running in the opposite direction. Reversing of the leads will switch the direction of the gel run, however, this will cause the current to flow from the anode to the cathode. The cathode electrode is made of stainless steel with platinum coating, and the anode electrode is made of platinum wire. Flow of electrons from the anode to the cathode will result in rusting of the steel core. On the other hand, when the leads are connected properly, the electrons flow from the cathode to the anode and the recipient of the electrons is the platinum wire that does not rust.

Note: When the Bolt gel cassette is inserted properly into the Bolt Mini Gel Tank or Mini Gel Tank, the lettering (gel type, SKU and expiration date) printed on the gel cassette reads from left to right (please see Page 11 of the manual (https://tools.thermofisher.com/content/sfs/manuals/mini_gel_tank_man.pdf).

I ran my Bolt Bis-Tris Plus gel using the Mini Gel Tank; the run was faster than normal and the resolution was very poor. What happened?

Here are possible causes and solutions:

1) Buffers are too concentrated or incorrect: Check buffer recipe; dilute or remake if necessary
2) Current is set at a higher limit: Decrease current to recommended running conditions.

I am running a Bolt Bis-Tris Plus gel using the Mini Gel Tank and the current reading on the power supply is very low (close to zero). What is the problem?

Here are possible causes and solutions:

1) Tape left on the bottom of the cassette: Remove tape from bottom of cassette.
2) Connection to power supply not complete: Check all connections with a voltmeter for conductance.
3) Insufficient buffer level: Make sure there is sufficient buffer in the electrophoresis tank to cover the wells of the gel.

I am running a Bolt Bis-Tris Plus gel and the run is taking longer than usual. What is the problem?

Here are possible causes and solutions:

1) Buffers are too dilute: Check buffer recipe; remake if necessary.
2) Buffer chamber is leaking: Make sure the cassette clamp is firmly seated, the gaskets are in place and the cassette clamp is locked.
3) Current is set too low: Set correct current

I am running a Bolt Bis-Tris Plus gel, and only one gel is running while the other isn't. What is the issue?

Here are some likely reasons and remedies:

*Leaking of buffer from the inside of the tank to the outside. Leaking is most likely due to inappropriate removal of cassettes, which can lead to the gasket not sitting properly and loss of a good seal. We recommend pushing the gasket down with the thumb.
*Not covering the wells completely with the buffer. We recommend filling the tank up to the electrodes prior to inserting the gel cassettes. Complete filling of the tank not only prevent leaks from the inside to the outside but also acts as a heat sink.
*Not removing the tape from one of the gel cassettes. Make sure that the tape is removed from both gel cassettes before inserting them into the tank.
*Not placing the gel cassette all the way to the bottom of the tank prior to clamping it (this will create a bypass of the current at the bottom, so the gel will run slower or not run at all).

I am trying to load my Bolt Bis-Tris Plus gel but am unable to see the sample wells. What should I do?

We recommend increasing the contrast between the sample well and the rest of the gel by marking the cassette at the bottom of the wells with a marker pen prior to placing the cassette in the electrophoresis tank.

When I tried to remove the comb from my Bolt Bis-Tris Plus gel, I noticed the presence of extra acrylamide. What is the purpose?

Due to the current wedge-well design of our Bolt gels, excess polyacrylamide (likely to be present in the area of the comb) is a consequence of our efforts to ensure that all wells are completely formed. Most of this excess material can be removed by slowly pulling the comb straight up out of the gel wells, turning the gel upside down, and then gently flicking the excess gel into a trash receptacle. Efforts are underway to reduce this excess.

The protein bands in some of my gel lanes are irregular or wavy? What would have caused this problem?

This could be due to:

*Debris in the well
*High salt in the sample (make sure that the salt concentration does not exceed 50-100 mM)
*Running buffer issue
*Gel casting error

I am seeing a very wavy and uneven dye front with my samples. Can you please help me troubleshoot?

This could be due to a gel polymerization issue combined with incorrect sample preparation (final sample dilution less than 1X). Please try a different lot of the same gel and make sure that the sample is correctly prepared.

I am seeing a faint, artifact doublet band at ~60 kDa in all my lanes. This band seems to be getting darker the longer I stain the gel. What could be causing this?

Possible cause:

*Excess reducing agent (beta-mercaptoethanol)
*Skin protein contaminants (keratin)

Remedy:

*The addition of iodoacetamide to the equilibration buffer just before applying the sample to the gel has been shown to eliminate these artifact bands.
*Use new electrophoretic solutions and wear gloves when handling and loading the gel. This issue is more common when highly sensitive stains are used.

I loaded different protein samples in each well but I see the same protein band in several neighboring lanes. What could have happened?

Possible cause:

*Carry-over contamination of sample from one well into neighboring wells due to loading error
*Contaminated running buffer
*Gel casting error: malformed wells

Remedy:

*Use a gel loading tip to load wells
*Reduce the sample volume
*Do not delay while loading wells
*Do not delay after the run, as proteins can diffuse horizontally; a full well left next to an empty well would eventually contaminate the empty well over time.

My protein bands appear to be skewed or distorted. What is the problem?

Possible cause:

*Poor polymerization around sample wells
*High salt concentration in sample
*Uneven gel interface
*Excessive pressure applied to the gel plates when the gel is placed into the clamp assembly
*Uneven heating of the gel
*Insoluble material in the gel or inconsistent pore size throughout the gel
*Air bubble during the run

Remedy:

*Remove excess salt/other material by dialysis, Sephadex G-25 or any other desalting column or using an Amicon concentrator.
*Either use a cooled apparatus or reduce the current at which electrophoresis is performed.

I ran my reduced protein samples under denaturing conditions and am seeing doublet protein bands when I expect to see single bands. Why is this happening?

A portion of the protein sample may have re-oxidized during the run, or may not have been fully reduced prior to the run. We recommend preparing fresh sample solution using fresh beta-mercaptoethanol or dithiothreitol (DTT). For NuPAGE gels, we recommend adding antioxidant to the running buffer.

My gel seems to be lifting off the cassette. What could be causing this?

Gel lifting off the cassette can be caused by:

*Expired gels that are degrading
*Improper storage of gels
*Too much heat accumulating during the electrophoresis run due to excessive current
*Insufficient polymerization of the polyacrylamide

I am seeing a faint shadow, or "ghost" band below a normal and expected protein band? What could be the potential issue?

Ghost bands are usually attributed to a slight lifting of the gel from the cassette, which results in the trickling down of some sample beyond its normal migration point. It then accumulates and appears as a faint second band.

My protein bands in the outer lanes of the gel show a "smiling" effect. Can you please help me troubleshoot?

"Smiling" bands may be the result of the acrylamide in the gel breaking down, leaving less of a matrix for the proteins to migrate. We recommend checking to ensure that the gels have not been used past their expiration date.

I see dumbbell or barbell shaped bands after protein electrophoresis. What could be causing this?

Barbell shaped bands are a result of loading too large of a sample volume. When a large sample volume is loaded, part of the sample tends to diffuse to the sides of the wells. When the run begins and the sample moves through the stacking portion of the gel, the sample will incompletely stack causing a slight retardation of the portion of the sample that diffused to the sides of the wells. This effect may be intensified for larger proteins, whose migration is more impeded in the low concentration acrylamide of the stacking gel. To alleviate the problem, we recommend concentrating the protein and loading a smaller volume. This gives a "thinner" starting zone.

Why do I get streaking forward or "frowning" of one of my samples on my protein gel?

Here are possible causes and solutions:

1) Sample overload: Do not overload samples
2) Addition of reducing agent that is not fresh: Reduce samples right before loading and do not use samples that have been stored in reducing agent
3)Re-oxidation of the protein during the run: Add antioxidant to the running buffer if you are running NuPAGE gels
4) Presence of highly hydrophobic regions where the protein can exclude SDS: Load the sample with 2X sample buffer instead of 1X sample buffer
5) Excess salt in the sample: Precipitate and reconstitute in lower salt buffer
6) Not enough SDS in the sample: Add SDS to the upper buffer chamber (try 0.1%, 0.2%, 0.3% and 0.4% SDS)

How are Bolt Bis-Tris Plus gels different from NuPAGE Bis-Tris gels?

While they are both Bis-Tris based gels, the chemistries are very different as Bolt Bis-Tris Plus gels have been optimized for western blotting. Another key difference is the enabling wedge well design for Bolt Bis-Tris Plus gels, which allows larger sample volume load.

What are the main advantages of NuPAGE gels over Invitrogen Tris-Glycine gels?

NuPAGE gels have the following advantages over Tris-Glycine gels:

*Higher stability and longer shelf life: NuPAGE Bis-Tris gels and NuPAGE Tris-Acetate gels have a lower operating pH (pH 7 for NuPAGE Bis-Tris gels and pH 8.1 for NuPAGE Tris-Acetate gels) than Invitrogen Tris-Glycine gels (pH 9.5). At basic pH, polyacrylamide hydrolyzes to polyacrylic acid and ammonia whereas at neutral pH, this hydrolysis is slower. Hence, NuPAGE gels have higher stability and longer shelf life than Invitrogen Tris-Glycine gels (12 months at 4-25 degrees C for NuPAGE Bis-Tris gels and 8 months at 4 degrees C for NuPAGE Tris-Acetate gels vs 4-8 weeks at 4 degrees C for Tris-Glycine gels).

*Better resolution of proteins due to:

- Reduced undesired chemical modifications: Free acrylamide alkylates proteins at basic pH (8.5 to 9.0). It targets sulfhydryl cysteines and amine groups at the N-terminus and on lysines. This modification does not happen at pH below 8. Hence, proteins run on NuPAGE gels undergo fewer of these undesired chemical modifications than those run on Tris-Glycine gels.

- Reduced hydrolysis of proteins: Heating of Tris-Glycine sample buffer (pH 6.8) results in a drop in pH, causing Asp-Pro cleavage of proteins. High temperature and longer duration of heating/boiling increase the rate of this cleavage resulting in multiple peptide bands of decreased intensity. At 100 degrees C, the pH drops as low as pH 4.3. On the other hand, NuPAGE LDS sample buffer (pH 8.5) drops to pH 8.1 when heated to 70 degrees C, avoiding this cleavage.

*Faster run times: 35-50 min for NuPAGE Bis-Tris gels and 1 hour for NuPAGE Tris-Acetate gels vs 90 min for Tris-Glycine gels

How does the operating pH for Tris-Glycine gels differ from that for NuPAGE Bis Tris and NuPAGE Tris-Acetate gels?

The operating pH for Tris-Glycine gels is 9.5; the operating pH for NuPAGE Bis-Tris gels is 7 and for NuPAGE Tris-Acetate gels is 8.1.

Which pre-stained protein standards are compatible with Bolt Bis-Tris Plus gels?

SeeBlue prestained standard, SeeBlue Plus 2 prestained standard and Invitrogen Sharp prestained standard are compatible with Bolt Bis-Tris Plus gels. Their migration is the same as that in NuPAGE gels with the corresponding buffer.

Do you offer Bolt Bis-Tris Plus gels in smaller packs (<10 gels/box)?

No. We only offer one size of 10 gels per box.

Should I use the NuPAGE Antioxidant when I run reduced protein samples in Bolt Bis-Tris Plus gels?

It is not necessary to use the Antioxidant while running reduced samples in Bolt Bis-Tris Plus gels but wouldn't hurt to use it.

What is the difference between your original and new Bolt Bis-Tris Plus gels?

The main difference is in the dimensions of the gels. Our New Bolt Bis-Tris Plus gels (Cat. No. NWxxxxxBOX) have the following dimensions:

*Cassette: 10 cm x 10 cm
*Gel: 8 cm x 8 cm

The new Bolt Bis-Tris Plus gels can be run using the new Mini Gel Tank (Cat. No. A25977), or the XCell SureLock Mini-Cell. To run these gels using the Bolt Mini Gel Tank (discontinued as of December 31, 2014), upgrading of the tank is necessary by replacing the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732) (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gel-electrophoresis-chamber-systems/mini-gel-tank/resources-upgrading-bolt-mini-gel-tank.html). Instructions for replacement of the cam handles can be found on Page 20 of the manual or in this video (https://www.youtube.com/watch?v=1FtiX8Skllw).

Our Original Bolt Bis-Tris Plus gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) had slightly different dimensions:

*Cassette: 10 cm x 10.5 cm
*Gel: 8 cm x 8.3 cm

Our Original Bolt Bis-Tris Plus Mini gels can only be run in the Bolt Mini gel tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).

What are the dimensions of Bolt Bis-Tris Plus gels?

Our New Bolt Bis-Tris Plus gels (Cat. No. NWxxxxxBOX) have the following dimensions:

*Cassette: 10 cm x 10 cm
*Gel: 8 cm x 8 cm

Our Original Bolt Bis-Tris Plus gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) had slightly different dimensions:

*Cassette: 10 cm x 10.5 cm
*Gel: 8 cm x 8.3 cm

What is the thickness of Bolt Bis-Tris Plus gels?

Bolt Bis-Tris Plus gels have a 1 mm thickness.

What is the percentage of the stacking gel in Bolt Bis-Tris Plus gels?

Bolt Bis-Tris Plus gels contain a 4% stacking gel.

What is the shelf life of Bolt Bis-Tris Plus gels?

Bolt Bis-Tris Plus gels have a shelf life of 16 months when stored at 4-25 degrees C.

How should I store my Bolt Bis-Tris Plus gels?

We recommend storing them at 4-25 degrees C. They should not be frozen.

What is the formulation of Bolt Bis-Tris Plus gels?

Bolt Bis-Tris Plus gels have a proprietary acrylamide/bisacrylamide formulation that is based on the NuPAGE Bis-Tris gel chemistry but different from it.

Can I run Mini gels with 10 cm gel cassettes using a Bolt Mini Gel Tank?

To run Mini gels with 10 cm gel cassettes using a Bolt Mini Gel Tank (without replacement of 10.5 cm cassette clamp cam handles with 10 cm cassette clamp cam handles), please use the instructions provided on Page 22 of the manual (https://tools.thermofisher.com/content/sfs/manuals/mini_gel_tank_man.pdf).

Note: For optimal results, to run 10 cm cassette Mini gels with a Bolt Mini Gel Tank, one should replace the black 10.5 cm cassette clamp cam handles on the Bolt Mini Gel Tank with gray 10 cm cassette clamp cam handles (Cat. No. A26732). Instructions for replacement of the cam handles can be found on Page 20 of the manual (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gel-electrophoresis-chamber-systems/mini-gel-tank/resources-upgrading-bolt-mini-gel-tank.html) or in this video (https://www.youtube.com/watch?v=1FtiX8Skllw).

Additional resources can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gel-electrophoresis-chamber-systems/mini-gel-tank/resources-upgrading-bolt-mini-gel-tank.html).

How do you recommend transferring Midi gels?

Midi gels can be transferred using:

*iBlot Dry Blotting System in conjunction with Transfer Stacks
*Invitrogen Semi-Dry Blotter for simultaneous transfer of up to 2 Midi-gels
*Thermo Scientific Power Blotter for simultaneous transfer of up to 2 Midi gels
*Thermo Scientific G2 Fast Blotter (will be discontinued as soon as we exhaust current inventory).

Will NP-40 affect the migration of my protein samples?

All detergents, or even phospholipids in cell extracts, will form mixed micelles with SDS and migrate down into the gel. They can also interfere with the SDS:protein binding equilibrium. Most of the non-ionic detergents, including NP-40, are the worst at interfering with SDS-PAGE. The rule of thumb is to keep the ratio of SDS to lipid or other detergent at 10:1 or greater to minimize these effects.

Do your Invitrogen protein gels contain any carbohydrates and are they suitable for carbohydrate analysis?

All Invitrogen protein gels contain sucrose as a density-adjusting agent to facilitate pouring of the gel. Protein samples run on Invitrogen gels would be contaminated with large amounts of sucrose. Thus, Invitrogen gels are not recommended for this application.

What is the material used for making your Invitrogen precast gel plastic cassettes?

The cassettes are made of a styrene copolymer.

Can I recycle your Invitrogen precast gel plastic cassettes?

We do not recommend recycling our plastic cassettes because they have a chemical coating on them that may produce toxic fumes when melted and potentially cause contamination.

What is the difference between Invitrogen Mini and Midi gel formats?

Midi gels are wider than Mini gels and hence have a larger number of wells to accommodate additional samples in one gel. An experiment from a Mini gel can be easily scaled-up to a Midi gel of the same gel chemistry.

Midi gels:
*NuPAGE Bis-Tris, NuPAGE Tris-Acetate, & Invitrogen Tris-Glycine: Gel dimensions are 13cm x 8.3cm and Cassette dimensions are 15cm x 10.3cm.

Mini gels:
*NuPAGE Bis-Tris, NuPAGE Tris-Acetate, & Invitrogen Tris-Glycine: Gel dimensions are 8cm x 8cm and Cassette dimensions are 10cm x 10cm.
*New Bolt Bis-Tris Plus (Cat. No. NWxxxxxBOX): Gel dimensions are 8cm x 8.3cm and Cassette Dimensions are 10cm x10cm.
*Original Bolt Bis-Tris Plus (Cat. No. BGxxxxxBOX): Gel dimensions are 8cm x 8.3cm and Cassette Dimensions are 10cm x 10.5cm.

Do you offer Bolt Bis-Tris Plus gels in the Midi format?

Bolt Bis-Tris Plus gels are not available in the Midi format.

What are the dimensions of your precast protein gels?

All of our Invitrogen precast protein gels (NuPAGE gels, Bolt Bis-Tris Plus gels, and Novex gels) are available in Mini format. Our Mini gel dimensions are 8 cm x 8 cm and the cassette dimensions are 10 cm x 10 cm.

Our NuPAGE Bis-Tris, NuPAGE Tris-Acetate, and Novex Tris-Glycine Plus gels are also available in the wider Midi format. Our Midi gel dimensions are 8 cm x 13 cm and the cassette dimensions are 10 cm x 15 cm.

Are your precast protein gels available in Mini and Midi formats?

All our Invitrogen protein gels are available in Mini format. Certain gel chemistries (NuPAGE Bis-Tris, NuPAGE Tris-Acetate, and Invitrogen Tris-Glycine gels) are also available in the wide Midi format.

Note that Bolt Bis-Tris gels are not available in the Midi format and our Thermo Scientific Precise precast gels are only available in Mini format.

When running two protein gels, do I need to double the voltage?

If you are running the gels at constant voltage, you do not need to increase the voltage regardless of the number of gels. However, the resulting current and wattage observed will multiply linearly with the number of gels. Keep in mind that the expected total current for your gels should not exceed the current limit of the power supply, or else the current will plateau and the run will slow down. (For example: Recommended constant voltage for running a NuPAGE Bis-Tris gel with MES Buffer is 200 V, with a starting current of 110-125 mA/gel and end current of 70-80 mA/gel. If the power supply has a current limit of 500 mA, the maximum number of NuPAGE Bis-Tris gels that can be run at one time with full power is 500 mA/125 mA = 4 gels. Any additional gels will decrease the current per gel and increase the run time.

Can I run reduced and non-reduced protein samples on the same gel?

We do not recommend running reduced and non-reduced protein samples on the same gel, especially in adjacent lanes, since the reducing agent may have a carry-over effect on the non-reduced samples if they are in close proximity.

Can I store my reduced protein samples for later use?

We do not recommend storing reduced protein samples for long periods of time even if they are frozen because reoxidation of the sample may happen during storage, causing inconsistent results.

What is the ratio of acrylamide:bisacrylamide and percentage of cross-linker in your Invitrogen precast gels?

*Tris-Glycine gels (except 4% Tris-Glycine gels) have a 34.5:1 Acrylamide:bisacrylamide and 2.6% Crosslinker.

*4% Tris-Glycine gels have a 76:1 ratio Acrylamide:bisacrylamide and 1.3% Crosslinker.

What is the percentage of the stacking gel in your Invitrogen precast protein gels?

The percentage of the stacking gel is 4% in most of our gels including the Bolt Bis-Tris Plus gels. The NuPAGE Tris-Acetate gels contain a 3.2% stacking gel.

Do your Invitrogen precast protein gels contain a stacking gel?

Our Invitrogen precast protein gels contain a stacking gel that is ~8 to 9 mm long (it ends right above the first ridge on the cassette). The manufacturing method used results in an interface between the stacking and resolving gels that is not visually detectable.

What are the recommended sample loading volumes and protein loading amounts for your precast protein gels?

*Tris-Glycine and Invitrogen Tricine Mini gels: see here (http://tools.thermofisher.com/content/sfs/manuals/electrophoresisguide_man.pdf), Page 8

*NuPAGE Tris-Acetate and NuPAGE Bis-Tris Mini gels: see here (http://tools.thermofisher.com/content/sfs/manuals/nupage_tech_man.pdf), Page 10

*Bolt Bis-Tris Plus Mini gels: see here (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/bolt-bis-tris-gels.html)

*Thermo Scientific Precise Tris-HEPES gels: see here (https://tools.thermofisher.com/content/sfs/manuals/MAN0011499_Precise_Protein_Gels_UG.pdf), Page 1

*Midi gels (Invitrogen Tris-Glycine, NuPAGE Bis-Tris and NuPAGE Tris-Acetate): see here (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/novex_midigel_man.pdf), Page 4

*Thermo Scientific Precise Tris-Glycine gels: see here (https://tools.thermofisher.com/content/sfs/manuals/D25MAN0011814_Precise_TrisGlycine_Gels_UG.pdf), Page 1

Do your precast protein gels contain SDS?

Our precast protein gels do not contain SDS but they can be run under denaturing conditions when used with the appropriate denaturing running buffer.
Note: NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels, and Thermo Scientific Precise Tris-HEPES gels cannot be run under native conditions; they can only be run under denaturing conditions.

*Invitrogen Tris-Glycine gels: For Native electrophoresis, use Invitrogen Tris-Glycine Native Running Buffer. For Denaturing electrophoresis, use Invitrogen Tris-Glycine SDS Running Buffer

*NuPAGE Tris-Acetate gels: For Native electrophoresis, use Invitrogen Tris-Glycine Native Running Buffer. For Denaturing electrophoresis, use NuPAGE Tris-Acetate SDS Running Buffer

*NuPAGE Bis-Tris gels: For Denaturing electrophoresis, use NuPAGE MOPS-SDS Running Buffer or NuPAGE MES-SDS Running Buffer

*Bolt Bis-Tris Plus gels: For Denaturing electrophoresis, use Bolt MOPS SDS Running Buffer or Bolt MES SDS Running Buffer

*Thermo Scientific Precise Tris-Glycine gels: For Native electrophoresis, use Tris-Glycine SDS Running Buffer without SDS added. For Denaturing electrophoresis, use Tris-Glycine SDS Running Buffer.

*Thermo Scientific Precise Tris-HEPES gels: For Denaturing electrophoresis, use Tris-HEPES SDS Running Buffer.