Learn More
Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications.
$256.00 - $261.00
Specifications
| For Use With (Equipment) | Mini Gel Tank, XCell SureLock Mini-Cell |
|---|---|
| Gel Percentage | 10% |
| Gel Size | Mini |
| GelThickness | 1.0 mm |
| Gel Type | Bis-Tris Plus |
| Catalog Number | Mfr. No. | Wells | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Wells | Price | Quantity | |||||
NW00100BOX
|
Invitrogen™
NW00100BOX |
10-well |
Each for $256.00
|
|
|||||
NW00102BOX
|
Invitrogen™
NW00102BOX |
12-well |
Each for $256.00
|
|
|||||
NW00105BOX
|
Invitrogen™
NW00105BOX |
15-well |
Each for $256.00
|
|
|||||
NW00107BOX
|
Invitrogen™
NW00107BOX |
17-well |
Each for $261.00
|
|
|||||
Description
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt Bis-Tris Plus gels to obtain optimal results for day-to-day protein separation needs.
Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES or MOPS SDS running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.
• Optimized run conditions—separate proteins using constant voltage in approximately 20 minutes
• Better protein integrity—neutral pH sample preparation process preserves proteins
• Improved protein separation—offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than tris-glycine based gels
• Wedge-shaped wells—easier sample loading and up to two times higher sample volume than other 1.0mm-thick gels
Specifications
| Mini Gel Tank, XCell SureLock Mini-Cell | |
| Mini | |
| Bis-Tris Plus | |
| 3.5 to 260 kDa | |
| Molecular Weight | |
| 10 gels (1 box) | |
| Store at 4–25°C. Do not freeze. | |
| 8 cm |
| 10% | |
| 1.0 mm | |
| Denaturing | |
| Denaturing | |
| Bolt, WedgeWell | |
| Room Temperature | |
| 8 cm |