missing translation for 'onlineSavingsMsg'
Learn More

BOC-S-trityl-L-cysteine, 99%, Thermo Scientific™

$81.85 - $149.56

Chemical Identifiers

CAS 21947-98-8
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.592
MDL Number MFCD00038251
InChI Key JDTOWOURWBDELG-QHCPKHFHSA-N
Synonym boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
PubChem CID 11167161
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC399060010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399060050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

21947-98-8
463.592
JDTOWOURWBDELG-QHCPKHFHSA-N
11167161
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
C27H29NO4S
MFCD00038251
boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid

Specifications

21947-98-8
C27H29NO4S
boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
463.592
463.6
99%
MFCD00038251
JDTOWOURWBDELG-QHCPKHFHSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
11167161
BOC-S-trityl-L-cysteine
Videos
SDS
Documents

Documents

RUO – Research Use Only