missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BMS 986187, >98%

Catalog No. 598350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
598350 50 mg
598310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 598350 Supplier Tocris Bioscience™ Supplier No. 5983/50
Only null left

Potent δ-opioid receptor positive allosteric modulator

Potent δ-opioid receptor positive allosteric modulator (EC50 = 30 nM). Exhibits 100-fold selectivity for δ over μ-opioid receptor. Enhances affinity of [Leu5]-Enkephalin (Cat.No. 1889), SNC 80 (Cat.No. 0764) and TAN 67 (Cat.No. 0921) for the δ-opioid receptor.

Chemical Identifiers

CAS 684238-37-7
Molecular Formula C31H34O4

Specifications

Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Formula Weight 470.61
Purity >98%
Molecular Weight (g/mol) 470.609
Chemical Name or Material 3,4,5,6,7,9-Hexahydro-3,3,6,6-tetramethyl-9-[4-[(2-methylphenyl)methoxy]phenyl]-1H-xanthene-1,8(2H)-
Recommended Storage Store at 4°C