missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ BMS 688521
Description
BMS 688521 is a potent inhibitor of the LFA-1/ICAM-1 interaction (IC50 values are 2.5 and 78 nM in HUVEC adhesion and mouse specific adhesion assays respectively). Inhibits eosinophil accumulation in the lungs in a mouse allergic eosinophilic lung inflammation model.
Specifications
Specifications
| Chemical Name or Material | 6-[(5S,9R)-9-(4-Cyanophenyl)-3-(3,5-dichlorophenyl)-1-methyl-2,4-dioxo-1,3,7-triazaspiro[4,4]non-7-yl]-3-pyridinecarboxylic acid |
| CAS | 893397-44-9 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Molecular Formula | C26H19Cl2N5O4 |
| Formula Weight | 536.37 |
| Purity | 98% |