missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BMPR1B (ALK6), GST Tag Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Invitrogen™ PV6236

 View more versions of this product

Catalog No. PV6236


Only null left
Explore available promotions

Description

Description

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant The gene accession number for BMPR1B (ALK6) is NP_001194.1.

BMPR1B (also known as ALK6, CDw293) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.
Specifications

Specifications

O00238
Kinase Assay
658
Human BMPR1B (ALK6), GST Tag
Purified
-80°C, Avoid Freeze/Thaw Cycles
Activin receptor-like kinase 6; Acvrlk6; AI385617; Alk6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1B receptor; BMP15; Bmpr1b; BMPR-1B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1B; bone morphogenetic protein receptor type IB; bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type 1B; bone morphogenetic protein receptor, type IB; CDw293; CFK-43a; serine/threonine receptor kinase; serine/threonine-protein kinase receptor R6; SKR6
BMPR1B
Protein
Human
GST-tag
-80°C, Avoid Freeze/Thaw Cycles
Baculovirus
Liquid
See Label
50mM tris HCl with 0.25mM DTT, 0.1mM PMSF, 25% glycerol, 10mM glutathione, 0.1mM EDTA, 150mM NaCl and no preservative; pH 7.5
66.7 kDa
7.5
100 μg
RUO
CDw293
149-end
Unconjugated
Recombinant
149-end
Serine/Threonine Kinase
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions