Learn More
Invitrogen™ Human BMPR1B (ALK6), GST Tag Recombinant Protein
Recombinant Protein
$3060.00 - $3060.00
Specifications
| Accession Number | O00238 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Gene ID (Entrez) | 658 |
| Molecular Weight (g/mol) | 66.7 kDa |
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant The gene accession number for BMPR1B (ALK6) is NP_001194.1.
BMPR1B (also known as ALK6, CDw293) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.Specifications
| O00238 | |
| Kinase Assay | |
| 66.7 kDa | |
| CDw293 | |
| Human | |
| GST-tag | |
| Liquid |
| See Label | |
| 658 | |
| Human BMPR1B (ALK6), GST Tag | |
| Unconjugated | |
| Recombinant | |
| Baculovirus |