Learn More
Thermo Scientific™ Imject™ Blue Carrier™ Protein
Easily prepare effective immunogens with the highly-soluble, mollusk-derived hemocyanin and complete hapten-carrier conjugation kit.
Supplier: Thermo Scientific™ 77130
Description
Thermo Scientific™ Blue Carrier Protein is a highly-soluble, mollusk-derived hemocyanin formulated in convenient package sizes and a complete kit for carbodiimide (EDC-mediated) hapten-carrier conjugation via amine and carboxyl groups.
Blue Carrier Protein is a purified preparation of Concholepas concholepas hemocyanin (CCH). The large protein exhibits most of the same immunogenic properties as the popular carrier protein, keyhole limpet hemocyanin (KLH). However, its signficantly better solubility provides greater flexibility in immunogen preparation protocols by allowing a broader range of buffer and pH conditions for coupling peptides, proteins and other haptens using classical crosslinkers such as glutaraldehyde, Sulfo-SMCC and EDC.Highlights:
Convenient packages – choose lyophilized Blue Carrier Protein or complete EDC conjugation kit
High-yield conjugation – each molecule of CCH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers
Validated quality – the immunogenic carrier protein is specially purified to ensure uniform chemical structure and solubility for consistent performance
Superior solubility – increased solubility compared to KLH allows Blue Carrier Protein to be used in a broader range of conjugation protocols to accommodate unusual or poorly soluble haptens
Highly immunogenic – provides essentially the same level of immunogenicity and phylogenetic distance as keyhole limpet hemocyanin (KLH)
Properties of Blue Carrier Protein:
- Alternative names: Concholepas concholepas hemocyanin, CCH
- Molecular weight: Whole molecule: ∽8,000,000; Subunits: ∽400,000 and ∽350,000
- Absorption: maxima 280nm, 350nm and 558nm
- Extinction coefficient (Abs. 1mg/mL solution): 1.4 at 280nm; 0.35 at 346nm
- A280/A350 Ratio: ∽4.0
- Copper/protein ratio: ∽16 Cu2+/protein molecule
- Isoelectric point: 6.0
- Solubility: Soluble in aqueous buffers pH 7.0 to 8.0
- Stability: Stable in aqueous buffers (∽pH 7.2 without Ca2+ or Mg2+)
Includes:
Kit contains lyophilized Blue Carrier Protein (in MES buffer), EDC crosslinker, buffers and desalting columns to easily prepare ready-to-use immunogens for injection
Specifications
| 4°C. Shipped on ice. | |
| Imject Blue Carrier Protein | |
| 100 mg | |
| Imject |
For Research Use Only. Not for use in diagnostic procedures.