missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Blue Carrier™ Protein

Catalog No. p-4527366
Change view
Click to view available options
Quantity:
100 mg
500 mg
Catalog No. Quantity
PI77130 100 mg
PI77660 500 mg
2 options

Catalog No. PI77130

Supplier: Thermo Scientific™ 77130

Only null left

Easily prepare effective immunogens with the highly-soluble, mollusk-derived hemocyanin and complete hapten-carrier conjugation kit.

Thermo Scientific™ Blue Carrier Protein is a highly-soluble, mollusk-derived hemocyanin formulated in convenient package sizes and a complete kit for carbodiimide (EDC-mediated) hapten-carrier conjugation via amine and carboxyl groups.

Blue Carrier Protein is a purified preparation of Concholepas concholepas hemocyanin (CCH). The large protein exhibits most of the same immunogenic properties as the popular carrier protein, keyhole limpet hemocyanin (KLH). However, its signficantly better solubility provides greater flexibility in immunogen preparation protocols by allowing a broader range of buffer and pH conditions for coupling peptides, proteins and other haptens using classical crosslinkers such as glutaraldehyde, Sulfo-SMCC and EDC.

Highlights:

  • Convenient packages – choose lyophilized Blue Carrier Protein or complete EDC conjugation kit

  • High-yield conjugation – each molecule of CCH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers

  • Validated quality – the immunogenic carrier protein is specially purified to ensure uniform chemical structure and solubility for consistent performance

  • Superior solubility – increased solubility compared to KLH allows Blue Carrier Protein to be used in a broader range of conjugation protocols to accommodate unusual or poorly soluble haptens

  • Highly immunogenic – provides essentially the same level of immunogenicity and phylogenetic distance as keyhole limpet hemocyanin (KLH)

Properties of Blue Carrier Protein:

  • Alternative names: Concholepas concholepas hemocyanin, CCH
  • Molecular weight: Whole molecule: ∽8,000,000; Subunits: ∽400,000 and ∽350,000
  • Absorption: maxima 280nm, 350nm and 558nm
  • Extinction coefficient (Abs. 1mg/mL solution): 1.4 at 280nm; 0.35 at 346nm
  • A280/A350 Ratio: ∽4.0
  • Copper/protein ratio: ∽16 Cu2+/protein molecule
  • Isoelectric point: 6.0
  • Solubility: Soluble in aqueous buffers pH 7.0 to 8.0
  • Stability: Stable in aqueous buffers (∽pH 7.2 without Ca2+ or Mg2+)

Includes:

Kit contains lyophilized Blue Carrier Protein (in MES buffer), EDC crosslinker, buffers and desalting columns to easily prepare ready-to-use immunogens for injection

Specifications

Product Line Imject
Product Type Imject Blue Carrier Protein
Content And Storage 4°C. Shipped on ice.
Quantity 100 mg

For Research Use Only. Not for use in diagnostic procedures.