missing translation for 'onlineSavingsMsg'
Learn More

Black PN, TCI America™

Catalog No. B156125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B156125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B156125G Supplier TCI America Supplier No. B156125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2519-30-4
Molecular Formula C28H17N5Na4O14S4
Molecular Weight (g/mol) 867.664
MDL Number MFCD00078980
InChI Key AGKKBBSOKGLVTM-YENKNCRGSA-J
Synonym brilliant black, brilliant black 1, black pn, unii-2wpr32u0cp, blue black bn, brilliant black a, food black 1, brilliant black bn, 2wpr32u0cp, 1743 black
PubChem CID 6321379
IUPAC Name tetrasodium;(6E)-4-acetamido-5-oxo-6-[[7-sulfonato-4-[(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]hydrazinylidene]naphthalene-1,7-disulfonate
SMILES CC(=O)NC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=C(C(=NNC3=C4C=C(C=CC4=C(C=C3)N=NC5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-])C2=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 867.66
Physical Form Crystalline Powder
Chemical Name or Material Black PN
EINECSNumber (5)-1123
RTECSNumber QJ5950000
TSCA No