missing translation for 'onlineSavingsMsg'
Learn More

bisphenol A, SPEX CertiPrep™

Catalog No. S509
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S509 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S509 Supplier SPEX CertiPrep Group LLC Supplier No. S509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Bisphenol A80-05-71000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 80-05-7 , 67-64-1
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00002366
InChI Key IISBACLAFKSPIT-UHFFFAOYSA-N
PubChem CID 6623
ChEBI CHEBI:33216
IUPAC Name 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Acetone
Quantity 1 mL
Chemical Name or Material Bisphenol A
DOTInformation Nonhazardous
Recommended Storage Freezer

Product intended for laboratory use only