missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Trioxide, 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B112025KG

 View more versions of this product

Catalog No. 18609128


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1304-76-3
2.5 kg
Reagent
Amber Glass Bottle
1
95%
0.001

Chemical Identifiers

1304-76-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.