missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Trioxide, 95%, Spectrum™ Chemical

Catalog No. p-7165143
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
2.5 kg
50 kg
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18609130 500 g Amber Glass Bottle
18609128 2.5 kg Amber Glass Bottle
18609129 12 kg Poly Pail
18609131 50 kg Fiber Drum
4 options

Catalog No. 18609130

Supplier: Spectrum Chemical Mfg Cor B1120500GM

Only null left

Chemical Identifiers

CAS 1304-76-3

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade Reagent
Loss on Drying 0.001
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.