missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99.975% (metals basis)

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

$143.15 - $767.90

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
Synonym Bismuth salicylate, basic, Salicylic acid bismuth basic salt
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4039418
View Documents
Thermo Scientific Chemicals
04039418
50 g
Each for $143.15
Only null left
 
AA4039430
View Documents
Thermo Scientific Chemicals
04039430
250 g
Each for $338.95
Only null left
 
AA40394A1
View Documents
Thermo Scientific Chemicals
040394A1
1 kg
Each for $767.90
Only null left
 
Description

Description

Used as Adhesives and sealant chemicals, flame retardants, pigments, propellants and blowing agents. It is also used as solvents. Other uses are in batteries, electrical and electronic products, explosive materials ink, toner, and colorant products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1304-76-3
465.96
TYIXMATWDRGMPF-UHFFFAOYSA-N
dibismuth(3+) trioxidandiide
Bi2O3
MFCD00003462
Bismuth salicylate, basic, Salicylic acid bismuth basic salt
[O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

ca 1890°C
Yellow
Powder
99.975% (Metals basis)
MFCD00003462
Insoluble in water. Soluble in acids.
[O--].[O--].[O--].[Bi+3].[Bi+3]
465.96
Odorless
Bismuth(III) oxide
817°C
1304-76-3
50 g
Bi2O3
Bismuth salicylate, basic, Salicylic acid bismuth basic salt
TYIXMATWDRGMPF-UHFFFAOYSA-N
dibismuth(3+) trioxidandiide
465.96
8.9
Videos
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

EINECSNumber : 215-134-7

RTECSNumber : EB2984460

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only