missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99.975% (metals basis)

Catalog No. aa40394a1
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
250 g
50 g
Catalog No. Quantity
AA40394A1 1 kg
AA4039418 50 g
AA4039430 250 g
3 options

Catalog No. AA40394A1

Supplier: Thermo Scientific Chemicals 040394A1

Only null left

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

Used as Adhesives and sealant chemicals, flame retardants, pigments, propellants and blowing agents. It is also used as solvents. Other uses are in batteries, electrical and electronic products, explosive materials ink, toner, and colorant products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
Synonym Bismuth salicylate, basic, Salicylic acid bismuth basic salt
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ca 1890°C
Melting Point 817°C
Color Yellow
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.975% (Metals basis)
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 465.96
Odor Odorless
Density 8.9
Chemical Name or Material Bismuth(III) oxide
Show More Show Less
Hazard Category H303: May be harmful if swallowed.
EINECSNumber 215-134-7
RTECSNumber EB2984460
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only