missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ BIS-Tris, ULTROL™ Grade, Calbiochem™,
A zwitterionic buffer that exhibits only a very small shift in dissociation constant with respect to temperature
Supplier: MilliporeSigma™ 3913351KG
Description
Suitable for fractionation of hemoglobinSpecifications
| zwitterionic buffer | |
| 1 kg | |
| ≥99.0% | |
| [bis(2-Hydroxyethyl)imino]-tris(hydroxymethyl)methane | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
| 81462 | |
| 209.2 |
| 6976-37-0 | |
| by titration | |
| C8H19NO5 | |
| Soluble in water | |
| C(CO)N(CCO)C(CO)(CO)CO | |
| 209.242 | |
| CHEBI:41250 | |
| ULTROL |
Chemical Identifiers
| 6976-37-0 | |
| 209.242 | |
| [bis(2-Hydroxyethyl)imino]-tris(hydroxymethyl)methane | |
| CHEBI:41250 | |
| C(CO)N(CCO)C(CO)(CO)CO |
| C8H19NO5 | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 81462 | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area.