missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BIS-Tris, ULTROL™ Grade, Calbiochem™,

Catalog No. 391335100GM
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
391335100GM 100 g
3913351KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 391335100GM Supplier MilliporeSigma™ Supplier No. 391335100GM

A zwitterionic buffer that exhibits only a very small shift in dissociation constant with respect to temperature

Suitable for fractionation of hemoglobin.

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym [bis(2-Hydroxyethyl)imino]-tris(hydroxymethyl)methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Quantity 100 g
Assay Percent Range ≥99.0%
Solubility Information Soluble in water
Formula Weight 209.2
Grade by titration
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area.