missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals BIS-TRIS propane, 0.2M buffer soln., pH 9.0
CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol
$167.34 - $263.03
Chemical Identifiers
| CAS | 64431-96-5 |
|---|---|
| Molecular Formula | C11H26N2O6 |
| Molecular Weight (g/mol) | 282.337 |
| MDL Number | MFCD00004689 |
| InChI Key | HHKZCCWKTZRCCL-UHFFFAOYSA-N |
| PubChem CID | 125132 |
| ChEBI | CHEBI:40947 |
| IUPAC Name | 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(CNC(CO)(CO)CO)CNC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63943AE
|
Thermo Scientific Chemicals
J63943AE |
100 mL |
Each for $167.34
|
|
|||||
|
AAJ63943AK
|
Thermo Scientific Chemicals
J63943AK |
250 mL |
Each for $263.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 64431-96-5 | |
| 282.337 | |
| HHKZCCWKTZRCCL-UHFFFAOYSA-N | |
| CHEBI:40947 | |
| C(CNC(CO)(CO)CO)CNC(CO)(CO)CO |
| C11H26N2O6 | |
| MFCD00004689 | |
| 125132 | |
| 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol |
Specifications
| 64431-96-5 | |
| 9.0 | |
| 100 mL | |
| MFCD00004689 | |
| C(CNC(CO)(CO)CO)CNC(CO)(CO)CO | |
| 282.337 | |
| CHEBI:40947 |
| 0.2 M | |
| Liquid | |
| C11H26N2O6 | |
| HHKZCCWKTZRCCL-UHFFFAOYSA-N | |
| 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol | |
| 125132 | |
| BIS-TRIS propane buffer soln. |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only