missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS propane, 0.2M buffer soln., pH 8.5

Catalog No. AAJ62012AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62012AE 100 mL
AAJ62012AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62012AE Supplier Thermo Scientific Chemicals Supplier No. J62012AE

CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

Concentration 0.2 M
pH 8.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material BIS-TRIS propane buffer soln.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only