missing translation for 'onlineSavingsMsg'
Learn More

bis-Tris, ≥99%, MP Biomedicals™

Catalog No. p-4403876
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10103825 25 g
ICN10103880 100 g
ICN10103890 500 g
ICN10103891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10103825 Supplier MP Biomedicals Inc Supplier No. 0210103825
Only null left

Exhibits a very small shift in dissociation constant with respect to temperature

  • Zwitterionic buffer
  • Buffering agent
  • White Powder

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range >99%
Synonym 2, 2-bis(Hydroxymethyl)-2, 2', 2,2-bis(Hydroxymethyl)-2,2',2”-nitrilotriethanol
Formula Weight 209.2
Chemical Name or Material bis-Tris