missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bis-Tris, ≥99%, MP Biomedicals™
Description
- Zwitterionic buffer
- Buffering agent
- White Powder
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | >99% |
| Synonym | 2,2-bis(Hydroxymethyl)-2,2',2”-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2' |
| Formula Weight | 209.2 |
| Chemical Name or Material | bis-Tris |