missing translation for 'onlineSavingsMsg'
Learn More

Bis-Tris, ≥98% (Dry Basis), Ultrapure

Catalog No. AAJ1211236
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1211236 500 g Plastic bottle
AAJ1211222 100 g Plastic bottle
AAJ12112A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ1211236 Supplier Thermo Fisher Scientific Supplier No. J1211236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis[2 to Hydroxyethyl]imino to tris[hydroxymethyl]methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Melting Point 102°C to 105°C
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Packaging Plastic bottle
Beilstein 2205275
Formula Weight 209.24
Percent Purity ≥98%
Chemical Name or Material bis[2-Hydroxyethyl]imino-tris[hydroxymethyl]methane
Hazard Category H315-H319-H335
Precautionary Statement P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P390-P501c
EINECSNumber 230-237-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only