missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 7.0

Catalog No. AAJ63346AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63346AK 250 mL
AAJ63346AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63346AK Supplier Thermo Scientific Chemicals Supplier No. J63346AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Color Colorless
Concentration 0.2 M
pH 7.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material BIS-TRIS
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only