missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 6.0

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

Supplier:  Thermo Scientific Chemicals J62686AK

 View more versions of this product

Catalog No. AAJ62686AK


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

BIS-TRIS
Colorless
6.0
250 mL
Odorless
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
81462
6976-37-0
0.2 M
Liquid
C8H19NO5
MFCD00002853
C(CO)N(CCO)C(CO)(CO)CO
209.242
CHEBI:41250

Chemical Identifiers

6976-37-0
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
CHEBI:41250
C(CO)N(CCO)C(CO)(CO)CO
C8H19NO5
MFCD00002853
81462
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only