missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd

CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol

$538.60 - $538.60

Chemical Identifiers

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Molecular Weight (g/mol) 701.90
MDL Number MFCD00009593
InChI Key YNHIGQDRGKUECZ-UHFFFAOYSA-L
Synonym bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
PubChem CID 131664180
IUPAC Name ethane;methane;palladium(2+);triphenylphosphane;dichloride
SMILES [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC197320050
SDS View Documents
Thermo Scientific Chemicals
197320050
5 g Glass bottle
Each for $538.60
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13965-03-2
701.90
YNHIGQDRGKUECZ-UHFFFAOYSA-L
131664180
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C36H30Cl2P2Pd
MFCD00009593
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
ethane;methane;palladium(2+);triphenylphosphane;dichloride

Specifications

13965-03-2
Yellow
Glass bottle
[(C6H5)3P]2PdCl2
5 g
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
YNHIGQDRGKUECZ-UHFFFAOYSA-L
ethane;methane;palladium(2+);triphenylphosphane;dichloride
131664180
15% Pd
Powder
260°C
Authentic
C36H30Cl2P2Pd
MFCD00009593
06,59; 07,95; 08,151; 17,111
Solubility in water: insoluble.
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
701.90
701.89
Analysis
Bis(triphenylphosphine)palladium(II) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

Warning

EINECSNumber : 237-744-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only