missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(triphenylphosphine)nickel(II) Dichloride 96.0+%, TCI America™
Supplier: TCI America B157110G
Specifications
| Bis(triphenylphosphine)nickel(II) Dichloride | |
| Black-Green | |
| MFCD00009592 MFCD00003043 MFCD20489348 | |
| bis triphenylphosphine dichloronickel ii | |
| Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 654.18 | |
| 654.18 | |
| Crystalline Powder |
| 14264-16-5 | |
| C36H30Cl2NiP2 | |
| 10 g | |
| VHQPRJDILRYZBG-UHFFFAOYSA-L | |
| dichloronickelbis(ylium); bis(triphenylphosphane) | |
| 131664342 | |
| ≥96.0% (T) |
Chemical Identifiers
| 14264-16-5 | |
| 654.18 | |
| VHQPRJDILRYZBG-UHFFFAOYSA-L | |
| 131664342 | |
| Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C36H30Cl2NiP2 | |
| MFCD00009592 MFCD00003043 MFCD20489348 | |
| bis triphenylphosphine dichloronickel ii | |
| dichloronickelbis(ylium); bis(triphenylphosphane) |
Safety and Handling
RTECSNumber : QR6170000
TSCA : Yes