missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)nickel(II)chloride, 98%

CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol

$100.52 - $100.52

Chemical Identifiers

CAS 14264-16-5
Molecular Formula C36H30Cl2NiP2
Molecular Weight (g/mol) 654.18
MDL Number MFCD00009592 MFCD00003043 MFCD20489348
InChI Key VHQPRJDILRYZBG-UHFFFAOYSA-L
Synonym bis triphenylphosphine dichloronickel ii
PubChem CID 131664342
IUPAC Name nickel(2+);triphenylphosphane;dihydrochloride
SMILES Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC217500100
SDS View Documents
Thermo Scientific Chemicals
217500100
10 g Glass bottle
Each for $100.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14264-16-5
654.18
VHQPRJDILRYZBG-UHFFFAOYSA-L
131664342
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C36H30Cl2NiP2
MFCD00009592 MFCD00003043 MFCD20489348
bis triphenylphosphine dichloronickel ii
nickel(2+);triphenylphosphane;dihydrochloride

Specifications

14264-16-5
Gray to Green
Authentic
Glass bottle
[(C6H5)3P]2NiCl2
10 g
bis triphenylphosphine dichloronickel ii
VHQPRJDILRYZBG-UHFFFAOYSA-L
nickel(2+);triphenylphosphane;dihydrochloride
131664342
98%
Bis(triphenylphosphine)nickel(II)chloride, 98%
>300.0°C
97.5% min.
98%
C36H30Cl2NiP2
MFCD00009592 MFCD00003043 MFCD20489348
02,110; 09,147; 11,165; 16,118
Solubility in water: insoluble
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
654.18
654.2
Crystals or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause cancer.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 238-154-8

RTECSNumber : QR6170000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only