missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)copper Tetrahydroborate 90.0+%, TCI America™

Catalog No. b11395g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11395G Supplier TCI America Supplier No. B11395G
Only null left

Chemical Identifiers

CAS 16903-61-0
Molecular Formula C36H34BCuP2
Molecular Weight (g/mol) 602.97
MDL Number MFCD00013219
InChI Key YVXRFTLDBYUBGJ-UHFFFAOYSA-N
Synonym bis triphenylphosphine copper tetrahydroborate, bis triphenylphosphine copper i borohydride, bis-triphenylphosphino-cuprous borohydride, tetrahydridoboratobis triphenylphosphine copper i, bis triphenylphosphine copper i tetrahydridoborate, bis triphenylphosphine copper tetrahydroborate,? pph3 2cubh4
PubChem CID 10908163
IUPAC Name λ¹-copper(1+) bis(triphenylphosphane) boranuide
SMILES [BH4-].[Cu+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 602.97
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(triphenylphosphine)copper Tetrahydroborate