accessibility menu, dialog, popup

UserName

BH4CuPPPhPhPhPhPhPh

CAS RN 16903-61-0

IUPAC Name: λ¹-copper(1+) bis(triphenylphosphane) boranuide
Synonyms: λ¹-copper(1+) bis(triphenylphosphine) boranuide
Molecular Weight (g/mol): 602.97
Molecular Formula: C36H34BCuP2
InChi Key: YVXRFTLDBYUBGJ-UHFFFAOYSA-N
SMILES: [BH4-].[Cu+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results