missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(trimethylsilyl) Sulfate 98.0+%, TCI America™
$154.02 - $415.84
Chemical Identifiers
| CAS | 18306-29-1 |
|---|---|
| Molecular Formula | C6H18O4SSi2 |
| Molecular Weight (g/mol) | 242.436 |
| MDL Number | MFCD00008268 |
| InChI Key | KRUQDZRWZXUUAD-UHFFFAOYSA-N |
| Synonym | bis trimethylsilyl sulfate, bistrimethylsilylsulfate, ditrimethylsilyl sulfate, sulfuric acid, tms, bis trimethylsilyl sulphate, silanol, trimethyl-, sulfate 2:1, acmc-209ekb, bis-trimethylsilyl-sulfate |
| PubChem CID | 87573 |
| IUPAC Name | bis(trimethylsilyl) sulfate |
| SMILES | C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C |
Chemical Identifiers
| 18306-29-1 | |
| 242.436 | |
| KRUQDZRWZXUUAD-UHFFFAOYSA-N | |
| 87573 | |
| C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C |
| C6H18O4SSi2 | |
| MFCD00008268 | |
| bis trimethylsilyl sulfate, bistrimethylsilylsulfate, ditrimethylsilyl sulfate, sulfuric acid, tms, bis trimethylsilyl sulphate, silanol, trimethyl-, sulfate 2:1, acmc-209ekb, bis-trimethylsilyl-sulfate | |
| bis(trimethylsilyl) sulfate |
Specifications
| 18306-29-1 | |
| White | |
| C6H18O4SSi2 | |
| 5 g | |
| bis trimethylsilyl sulfate, bistrimethylsilylsulfate, ditrimethylsilyl sulfate, sulfuric acid, tms, bis trimethylsilyl sulphate, silanol, trimethyl-, sulfate 2:1, acmc-209ekb, bis-trimethylsilyl-sulfate | |
| C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C | |
| 242.436 | |
| 242.44 | |
| Crystalline Powder |
| 46°C | |
| 90°C | |
| MFCD00008268 | |
| 2925 | |
| KRUQDZRWZXUUAD-UHFFFAOYSA-N | |
| bis(trimethylsilyl) sulfate | |
| 87573 | |
| ≥98.0% (T) | |
| Bis(trimethylsilyl) Sulfate |
Safety and Handling
TSCA : No