missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(trimethylsilyl) Sulfate 98.0+%, TCI America™
Supplier: TCI America B124525G
Specifications
| Bis(trimethylsilyl) Sulfate | |
| 46°C | |
| 90°C | |
| MFCD00008268 | |
| 2925 | |
| KRUQDZRWZXUUAD-UHFFFAOYSA-N | |
| bis(trimethylsilyl) sulfate | |
| 87573 | |
| ≥98.0% (T) |
| 18306-29-1 | |
| White | |
| C6H18O4SSi2 | |
| 25 g | |
| bis trimethylsilyl sulfate, bistrimethylsilylsulfate, ditrimethylsilyl sulfate, sulfuric acid, tms, bis trimethylsilyl sulphate, silanol, trimethyl-, sulfate 2:1, acmc-209ekb, bis-trimethylsilyl-sulfate | |
| C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C | |
| 242.436 | |
| 242.44 | |
| Crystalline Powder |
Chemical Identifiers
| 18306-29-1 | |
| 242.436 | |
| KRUQDZRWZXUUAD-UHFFFAOYSA-N | |
| 87573 | |
| C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C |
| C6H18O4SSi2 | |
| MFCD00008268 | |
| bis trimethylsilyl sulfate, bistrimethylsilylsulfate, ditrimethylsilyl sulfate, sulfuric acid, tms, bis trimethylsilyl sulphate, silanol, trimethyl-, sulfate 2:1, acmc-209ekb, bis-trimethylsilyl-sulfate | |
| bis(trimethylsilyl) sulfate |
Safety and Handling
TSCA : No