missing translation for 'onlineSavingsMsg'
Learn More

Bis(trifluoromethanesulfonyl)imide 99.0+%, TCI America™

Catalog No. b25415g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B25415G 5 g
B254125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B25415G Supplier TCI America Supplier No. B25415G
Only null left

Chemical Identifiers

CAS 82113-65-3
Molecular Formula C2HF6NO4S2
Molecular Weight (g/mol) 281.143
MDL Number MFCD00214154
InChI Key ZXMGHDIOOHOAAE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amine
PubChem CID 157857
IUPAC Name 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 56°C
Color White
Boiling Point 92°C
Quantity 5 g
UN Number 3261
Formula Weight 281.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trifluoromethanesulfonyl)imide
TSCA Yes
Recommended Storage Refrigerator