missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-o-tolylphosphine)palladium(II) Dichloride 90.0+%, TCI America™

Catalog No. p-7119432
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B20261G 1 g
B20265G 5 g
2 options

Catalog No. B20261G

Supplier: TCI America B20261G

Only null left

Chemical Identifiers

CAS 40691-33-6
Molecular Formula C42H42Cl2P2Pd
Molecular Weight (g/mol) 786.06
MDL Number MFCD00274659
InChI Key GMLMZLGOMMIIMA-UHFFFAOYSA-L
Synonym palladium,dichlorobis tris 2-methylphenyl phosphine
PubChem CID 90656943
IUPAC Name dichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
SMILES Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 786.07
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tri-o-tolylphosphine)palladium(II) Dichloride