missing translation for 'onlineSavingsMsg'
Learn More

Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%

Catalog No. p-3586404
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377960010 1 g Glass bottle
AC377960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC377960010 Supplier Thermo Scientific Chemicals Supplier No. 377960010

CAS: 40691-33-6 | C42H42Cl2P2Pd | 786.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40691-33-6
Molecular Formula C42H42Cl2P2Pd
Molecular Weight (g/mol) 786.06
MDL Number MFCD00274659
InChI Key GMLMZLGOMMIIMA-UHFFFAOYSA-L
Synonym palladium,dichlorobis tris 2-methylphenyl phosphine
PubChem CID 90656943
IUPAC Name palladium;tris(2-methylphenyl)phosphane;dichloride
SMILES Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1

Specifications

Melting Point 280°C
Color Yellow to Orange
Elemental Analysis 94% min. (ex Carbon)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 786.06
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Bis[tri(o-tolyl)phosphine]palladium(II) chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only