missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylthio)methane, 98+%

Catalog No. AAL0172306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0172306 5 g
AAL0172314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0172306 Supplier Thermo Scientific Chemicals Supplier No. L0172306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3561-67-9 | C13H12S2 | 232.359 g/mol

Acyl anion equivalent with some advantages over 1,3-dithiane for the preparation of vinyl sulfides. Bis(phenylthio)methane is used in the synthesis of the dihydroxy thioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acyl anion equivalent with some advantages over 1,3-dithiane for the preparation of vinyl sulfides. Bis(phenylthio)methane is used in the synthesis of the dihydroxy thioethers.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 3561-67-9
Molecular Formula C13H12S2
Molecular Weight (g/mol) 232.359
MDL Number MFCD00003067
InChI Key ZHUPZVIALZHGGP-UHFFFAOYSA-N
Synonym bis phenylthio methane, methylenebis thio bisbenzene, bis phenyithio methane, formaldehyde diphenyl mercaptal, 1,1'-methylenebis thio bisbenzene, phenylsulfanyl methyl sulfanyl benzene, 1-phenylthio methyl thio benzene, bis thiophenoxy methane
PubChem CID 77097
IUPAC Name phenylsulfanylmethylsulfanylbenzene
SMILES C1=CC=C(C=C1)SCSC2=CC=CC=C2

Specifications

Melting Point 36°C to 38°C
Boiling Point 194°C (8 mmHg)
Linear Formula C6H5SCH2SC6H5
Quantity 5 g
Beilstein 1874146
Solubility Information Insoluble in water.
Formula Weight 232.36
Percent Purity ≥98%
Chemical Name or Material Bis(phenylthio)methane
EINECSNumber 222-624-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only