missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylacetyl) Disulfide 98.0+%, TCI America™

Supplier:  TCI America B362325G

 View more versions of this product

Catalog No. B362325G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis(phenylacetyl) Disulfide
60°C
C16H14O2S2
25 g
IXGZXXBJSZISOO-UHFFFAOYSA-N
2-phenyl-1-[(2-phenylacetyl)disulfanyl]ethan-1-one
3378603
≥98.0% (HPLC)
15088-78-5
White
MFCD00513572
phenylacetyl disulfide, 2-phenylacetic dithioperoxyanhydride, phenylacetyl disulphide, bis phenylacetyl disulfide, bis phenylacetyl disulphide, 2-phenyl-1-2-phenylacetyl disulfanyl ethanone, bis phenylacetyl persulfide, phenylacetyl disulfide 5g, s-2-phenylacetyl sulfanyl 2-phenylethanethioate
O=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1
302.41
302.41
Crystalline Powder

Chemical Identifiers

15088-78-5
302.41
IXGZXXBJSZISOO-UHFFFAOYSA-N
3378603
O=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1
C16H14O2S2
MFCD00513572
phenylacetyl disulfide, 2-phenylacetic dithioperoxyanhydride, phenylacetyl disulphide, bis phenylacetyl disulfide, bis phenylacetyl disulphide, 2-phenyl-1-2-phenylacetyl disulfanyl ethanone, bis phenylacetyl persulfide, phenylacetyl disulfide 5g, s-2-phenylacetyl sulfanyl 2-phenylethanethioate
2-phenyl-1-[(2-phenylacetyl)disulfanyl]ethan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions