missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylacetyl) Disulfide 98.0+%, TCI America™

Catalog No. p-7123056
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36235G 5 g
B362325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36235G Supplier TCI America Supplier No. B36235G
Only null left

Chemical Identifiers

CAS 15088-78-5
Molecular Formula C16H14O2S2
Molecular Weight (g/mol) 302.41
MDL Number MFCD00513572
InChI Key IXGZXXBJSZISOO-UHFFFAOYSA-N
Synonym phenylacetyl disulfide, 2-phenylacetic dithioperoxyanhydride, phenylacetyl disulphide, bis phenylacetyl disulfide, bis phenylacetyl disulphide, 2-phenyl-1-2-phenylacetyl disulfanyl ethanone, bis phenylacetyl persulfide, phenylacetyl disulfide 5g, s-2-phenylacetyl sulfanyl 2-phenylethanethioate
PubChem CID 3378603
IUPAC Name 2-phenyl-1-[(2-phenylacetyl)disulfanyl]ethan-1-one
SMILES O=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1

Specifications

Melting Point 60°C
Color White
Quantity 5 g
Formula Weight 302.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(phenylacetyl) Disulfide