accessibility menu, dialog, popup

UserName

CAS RN 15088-78-5

IUPAC Name: 2-phenyl-1-[(2-phenylacetyl)disulfanyl]ethan-1-one
Synonyms: 2-phenyl-1-[(2-phenylacetyl)disulfanyl]ethanone
Molecular Weight (g/mol): 302.41
Molecular Formula: C16H14O2S2
InChi Key: IXGZXXBJSZISOO-UHFFFAOYSA-N
SMILES: O=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)