missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-toluenesulfonyl)methane, 97%

Catalog No. p-7029178
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2661703 1 g
AAH2661706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2661703 Supplier Thermo Scientific Chemicals Supplier No. H2661703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15310-28-8 | C15H16O4S2 | 324.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15310-28-8
Molecular Formula C15H16O4S2
Molecular Weight (g/mol) 324.41
MDL Number MFCD00032255
InChI Key XLOXYWLRAKCDQL-UHFFFAOYSA-N
Synonym ditosylmethane, bis-toluene-4-sulfonyl-methane, 1-methyl-4-4-methylbenzenesulfonyl methanesulfonyl benzene, 1,1'-methanediyldisulfonyl bis 4-methylbenzene, acmc-20allu, bis p-tolylsulfonyl methane, bis p-toluenesulfonyl methane, bis-toluene-4-sulfonyl methane, 1,1'-methylenedisulfonyl bis 4-methylbenzene, benzene,1,1'-methylenebis sulfonyl bis 4-methyl
PubChem CID 275690
IUPAC Name 1-methyl-4-[(4-methylphenyl)sulfonylmethylsulfonyl]benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 137°C to 139°C
Quantity 1 g
Formula Weight 324.42
Percent Purity 97%
Chemical Name or Material Bis(p-toluenesulfonyl)methane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only