missing translation for 'onlineSavingsMsg'
Learn More

Bis[bis[4-(diethylamino)phenyl]methyl] Ether 97.0+%, TCI America™

Catalog No. B26755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26755G Supplier TCI America Supplier No. B26755G

Chemical Identifiers

CAS 155050-06-9
Molecular Formula C42H58N4O
Molecular Weight (g/mol) 634.953
InChI Key DTWIRTBPOMOUFA-UHFFFAOYSA-N
Synonym Bis[4,4′C-bis(diethylamino)benzhydryl] Ether
PubChem CID 44629767
IUPAC Name 4-[bis[4-(diethylamino)phenyl]methoxy-[4-(diethylamino)phenyl]methyl]-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)C(C2=CC=C(C=C2)N(CC)CC)OC(C3=CC=C(C=C3)N(CC)CC)C4=CC=C(C=C4)N(CC)CC

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 634.95
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[bis[4-(diethylamino)phenyl]methyl] Ether