CAS RN 155050-06-9
IUPAC Name:
4-({bis[4-(diethylamino)phenyl]methoxy}[4-(diethylamino)phenyl]methyl)-N,N-diethylaniline
Synonyms:
4-({bis[4-(diethylamino)phenyl]methoxy}[4-(diethylamino)phenyl]methyl)-N,N-diethylaniline
Molecular Weight (g/mol):
634.95
Molecular Formula:
C42H58N4O
InChi Key:
DTWIRTBPOMOUFA-UHFFFAOYSA-N
SMILES:
CCN(CC)C1=CC=C(C=C1)C(OC(C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
1
–
1
of
1
results
1
Bis[bis[4-(diethylamino)phenyl]methyl] Ether 97.0+%, TCI America™
CAS: 155050-06-9 Molecular Formula: C42H58N4O Molecular Weight (g/mol): 634.953 InChI Key: DTWIRTBPOMOUFA-UHFFFAOYSA-N Synonym: Bis[4,4′C-bis(diethylamino)benzhydryl] Ether PubChem CID: 44629767 IUPAC Name: 4-[bis[4-(diethylamino)phenyl]methoxy-[4-(diethylamino)phenyl]methyl]-N,N-diethylaniline SMILES: CCN(CC)C1=CC=C(C=C1)C(C2=CC=C(C=C2)N(CC)CC)OC(C3=CC=C(C=C3)N(CC)CC)C4=CC=C(C=C4)N(CC)CC