missing translation for 'onlineSavingsMsg'
Learn More

Bis(9,9-dimethyl-9H-fluoren-2-yl)amine 98.0+%, TCI America™

Catalog No. p-7123677
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46641G 1 g
B46645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46641G Supplier TCI America Supplier No. B46641G
Only null left

Chemical Identifiers

CAS 500717-23-7
Molecular Formula C30H27N
Molecular Weight (g/mol) 401.553
MDL Number MFCD23135317
InChI Key LCSMGMWMTSWXDD-UHFFFAOYSA-N
PubChem CID 59254080
IUPAC Name N-(9,9-dimethylfluoren-2-yl)-9,9-dimethylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)NC4=CC5=C(C=C4)C6=CC=CC=C6C5(C)C)C

Specifications

Melting Point 184°C
Quantity 1 g
Formula Weight 401.55
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(9,9-dimethyl-9H-fluoren-2-yl)amine